Merck
  • Capillary gas chromatographic determination of methylglyoxal from serum of diabetic patients by precolumn derivatization with 1,2-diamonopropane.
Capillary gas chromatographic determination of methylglyoxal from serum of diabetic patients by precolumn derivatization with 1,2-diamonopropane.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-09-02)
M Y Khuhawar, L A Zardari, A J Laghari
ABSTRACT
Capillary gas chromatographic (GC) determination of methylglyoxal (MGo) was developed on the basis of precolumn derivatization with 1,2-diaminopropane (DAP) at pH 3. The elution was carried out on an HP-5 (30 m x 0.32 mm i.d.) connected with FID. The linear calibration curve was obtained for MGo within 0.09-1.04 microg/ml with detection limit of 40 ng/ml. Dimethylglyoxal (DMGo) also formed derivative with DAP and eluted and separated from MGo at column temperature 100 degrees C for 1 min with heating rate 30 degrees C/min up to 200 degrees C with run time 4.6 min. The nitrogen flow rate was 1.5 ml/min with split ratio of 10:1, v/v. MGo was determined from serum and urine of diabetics and healthy volunteers. The amounts of MGo from serum and urine of diabetic patients were 0.180-0.260 microg/ml and 0.170-0.250 microg/ml with relative standard deviation (R.S.D.) within 1-4% and 1-3%, respectively. The amounts of MGo from serum of healthy volunteers were 0.032-0.054 microg/ml with an R.S.D. of 1.5-3%. DMGo was not detected from the biological fluids and was used as an internal standard.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Acetone, analytical standard
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
(S)-(−)-1,2-Diaminopropane dihydrochloride, 99%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
(R)-(+)-1,2-Diaminopropane dihydrochloride, 99%
Sigma-Aldrich
1,2-Diaminopropane, 99%
Sigma-Aldrich
4-Heptanone, 98%
Sigma-Aldrich
(R)-3-Hydroxybutyric acid, ≥98.0% (T)