Merck
[Metabolism and toxicity of n-pentane and isopentane].
Nihon hoigaku zasshi = The Japanese journal of legal medicine (1991-04-01)
S Chiba, S Oshida
ABSTRACT
n-Pentane and isopentane have a wide range of use, for example, for cleaning precision machinery, extracting essence and oil, and as liquid fuel for now very popular disposable lighters. They are contained in liquefied petroleum gas and natural gas as trace constituents. In our present experiments, we studied the metabolism and toxicity of these n-pentane and isopentane metabolites. Male mice of ICR strain were exposed to about 5% n-pentane for one hour while the oxygen in the environmental air was maintained at about 20%. Then their blood and liver tissue were collected and analyzed by means of GC and GC-MS. The metabolites thus obtained were 2-pentanol, 3-pentanol and 2-pentanone. The same procedure was repeated with isopentane; 3-methyl-2-butanol, 2-methyl-2-butanol and 3-methyl-2-butanone were detected as the resultant metabolites. In the presence of the NADPH-generating system liver microsomes were made to react to the substrate of saturated n-pentane or isopentane aqueous solution at 37 degrees C for one hour. As a result, the same metabolites were produced as obtained in the exposure experiment. It was therefore suggested that n-pentane and isopentane were metabolized chiefly by liver microsomes. Male mice of ICR strain were fed with 80 mg/kg b.w. of phenobarbital for consecutive four days and exposed to n-pentane or isopentane for one hour. This resulted in an increase in the amount of 2-pentanol and 2-pentanone in the n-pentane inhalation and 2-methyl-2-butanol in the isopentane inhalation experiment. The toxicity of each metabolite was studied on cultured cells. The metabolites were individually mixed with HeLa S3 cell suspension, incubated for three days, and their concentration which inhibited the growth of cells by 50% (IGC 50) were compared. It was demonstrated as a result that the IGC 50 for any of the metabolites was lower than that for methanol, ethanol or acetone used as control.
Product Description
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%