- Glutathione-S-transferase-mediated methylthio displacement and turnover in the metabolism of pentachlorothioanisole by rats.
Glutathione-S-transferase-mediated methylthio displacement and turnover in the metabolism of pentachlorothioanisole by rats.
Xenobiotica; the fate of foreign compounds in biological systems (1991-05-01)
D J Mulford, J E Bakke, V J Feil
PMID1949892
ABSTRACT
1. The metabolism of pentachlorothioanisole to bis-(methylthio)tetrachlorobenzene was shown to involve turnover of about 50% of the original methylthio group. 2. Metabolites of the displaced methylthio group were methanesulphinic acid and sulphate in the urine, carbon dioxide in the expired air, and unidentified sulphur-containing compounds in the bile.