Merck
  • Effects of single and repeated treatment with antidepressants on apomorphine-induced yawning in the rat: the implication of alpha-1 adrenergic mechanisms in the D-2 receptor function.
Effects of single and repeated treatment with antidepressants on apomorphine-induced yawning in the rat: the implication of alpha-1 adrenergic mechanisms in the D-2 receptor function.
Psychopharmacology (1990-01-01)
A Delini-Stula, C Hunn
ABSTRACT
Acute (10 or 20 mg/kg IP) and subchronic (2 x 5 or 10 mg/kg IP daily for 7 days) effects of desipramine, imipramine, maprotiline, (+)- and (-)-oxaprotiline enantiomers as well as selective 5-HT-uptake inhibitors citalopram and ifoxetine on yawning, induced by low doses of apomorphine, were investigated in the rat. In addition, the effects of alpha-1 receptor agonist adrafinil and antagonist prazosin were also tested. After acute treatment, desipramine, the stereoselective NA-uptake inhibiting (+)-enantiomer of oxaprotiline, and the alpha-1 agonist adrafinil, markedly and significantly suppressed yawning. Prazosin, in contrast, clearly potentiated it. This potentiating effect was abolished by the pretreatment with (+)-oxaprotiline and adrafinil. Other drugs were inactive. After subchronic administration, yawning was antagonized by NA-uptake-inhibiting antidepressants, including imipramine and maprotiline. By comparison to the acute treatment, the inhibitory effects of desipramine and (+)-oxaprotiline were considerably enhanced. Neither selective 5-HT-uptake inhibitors nor (-)-oxaprotiline (levoprotiline) were active. Antidepressants therefore modulate the functional activity of D-2 receptors, activated by low doses of apomorphine, predominantly by the virtue of their noradrenergic enhancing properties. This modulatory effect appears to be mediated by alpha-1 adrenergic receptors.