Merck
  • Small quantities of carotenoid-rich tropical green leafy vegetables indigenous to Africa maintain vitamin A status in Mongolian gerbils ( Meriones unguiculatus).
Small quantities of carotenoid-rich tropical green leafy vegetables indigenous to Africa maintain vitamin A status in Mongolian gerbils ( Meriones unguiculatus).
The British journal of nutrition (2010-04-24)
Richard A Ejoh, Joseph T Dever, Jordan P Mills, Sherry A Tanumihardjo
ABSTRACT
Leafy vegetables are important sources of provitamin A carotenoids. Information on their ability to provide vitamin A is often misleading because of the methodology used to assess bioefficacy. Mongolian gerbils were used to evaluate the bioefficacy of provitamin A carotenoids in tropical leafy vegetables (i.e. Solanum nigrum, Moringa oleifera, Vernonia calvoana and Hibiscus cannabinus) that are indigenous to Africa. Gerbils (n 67) were vitamin A-depleted for 5 weeks. After a baseline kill (n 7), the gerbils were weight-matched and assigned to six treatment groups (n 10; four vegetable groups; negative and positive controls). For 4 weeks, the treatments included 35 nmol vitamin A (theoretical concentrations based on 100 % bioefficacy) in the form of vegetables or retinyl acetate. In addition to their diets, the control and vegetable groups received daily doses of oil, while the vitamin A group received retinyl acetate in oil matched to prior day intake. Serum and livers were analysed for vitamin A using HPLC. Serum retinol concentrations did not differ among groups, but total liver vitamin A of the vitamin A and vegetable groups were higher than that of the negative control group (P < 0.0001). Liver beta-carotene 15,15'-monooxygenase-1 expression levels were determined for two vegetable groups and were similar to the positive and negative controls. Conversion factors for the different leafy vegetables were between 1.9 and 2.3 microg beta-carotene equivalents to 1 microg retinol. Small quantities of these vegetables maintained vitamin A status in gerbils through efficient bioconversion of beta-carotene to retinol.
Product Description
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Supelco
Retinyl acetate, analytical standard
Retinol acetate, European Pharmacopoeia (EP) Reference Standard