Merck
  • Synthesis and biological evaluation of novel spiro 6-methoxytetralin-1,3'-pyrrolidine based organoselenocyanates against cadmium-induced oxidative and hepatic damage in mice.
Synthesis and biological evaluation of novel spiro 6-methoxytetralin-1,3'-pyrrolidine based organoselenocyanates against cadmium-induced oxidative and hepatic damage in mice.
European journal of medicinal chemistry (2010-05-12)
Ugir Hossain Sk, Arun K Sharma, Sulekha Ghosh, Sudin Bhattacharya
ABSTRACT
A series of novel organoselenocyanate compounds (6a-d) having spiro[tetralin-1,3'-pyrrolidine] moiety were synthesized. The compounds were evaluated for their inhibitory activity against cadmium- (Cd) induced toxicity in Swiss Albino mice. All the compounds (6a-d) inhibited the level of lipid peroxidation (LPO) and upregulated the activity of glutathione-S-transferase (GST), superoxide dismutase (SOD), catalase (CAT) and reduced glutathione (GSH) levels in treatment group in comparison to the untreated Cd control group. Serum transaminase activities, e.g., alanine aminotransferase (ALT) and aspartate aminotransferase (AST) were also significantly lowered in the compound-treated mice. Elongation of the alkyl chain length bearing the selenocyanate (SeCN) active group enhanced the potency of the compounds, 6d being the most active one (6d>6c>6b>6a).
Sigma-Aldrich
Potassium selenocyanate, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium selenocyanate, reagent grade, 97%