Merck
  • A fish-oil-rich diet reduces vascular oxidative stress in apoE(-/-) mice.
A fish-oil-rich diet reduces vascular oxidative stress in apoE(-/-) mice.
Free radical research (2010-06-10)
Kelly Casós, María C Zaragozá, Neven Zarkovic, Kamelija Zarkovic, Luka Andrisic, Manuel Portero-Otín, Daniel Cacabelos, María T Mitjavila
ABSTRACT
Oxidative stress contributes to lipid peroxidation and decreases nitric oxide (NO) bioavailability in atherosclerosis. While long-chain (n-3) polyunsaturated fatty acids (PUFA) are easily oxidized in vitro, they improve endothelial function. Hence, this study postulates that long-chain (n-3) PUFA decrease atherogenic oxidative stress in vivo. To test this, apoE(-/-) mice were fed a corn oil- or a fish oil (FO)-rich diet for 8, 14 or 20 weeks and parameters related to NO and superoxide (O(2)(.-)) plus markers of lipid peroxidation and protein oxidative damage in the aortic root were evaluated. The FO-rich diet increased NO production and endothelial NO synthase (NOS) expression and lowered inducible NOS, p22(phox) expression and O(2)(.-)production after 14 and 20 weeks of diet. Protein lipoxidative damage (including 4-hydroxynonenal) was decreased after a long-term FO-diet. This supports the hypothesis that a FO-rich diet could counteract atherogenic oxidative stress, showing beneficial effects of long-chain (n-3) PUFA.
Sigma-Aldrich
Fish oil from menhaden, crude source of omega-3 fatty acids