Merck
  • Allergic contact dermatitis to propyl gallate and pentylene glycol in an emollient cream.
Allergic contact dermatitis to propyl gallate and pentylene glycol in an emollient cream.
The Australasian journal of dermatology (2010-06-16)
Caterina Foti, Domenico Bonamonte, Nicoletta Cassano, Anna Conserva, Gino A Vena
ABSTRACT
A 62-year-old man, with a 20-year history of seborrhoeic dermatitis, presented with a worsening of his dermatitis. He had previously been demonstrated to be allergic to various topical corticosteroids, so he had been using an emollient cream (Sebclair), containing piroctone olamine and various anti-inflammatory substances, for 6 months, with good effect. Patch testing to the cream and its ingredients revealed positive reactions to both propyl gallate and pentylene glycol. A positive reaction to propylene glycol was also detected, whereas patch testing to butylene glycol was negative. Complete remission followed avoidance of the offending substances.
Sigma-Aldrich
1,5-Pentanediol, 96%
Sigma-Aldrich
1,5-Pentanediol, purum, ≥97.0% (GC)