Merck
  • Determination of aflatoxins B1, B2, G1, and G2 in olive oil, peanut oil, and sesame oil.
Determination of aflatoxins B1, B2, G1, and G2 in olive oil, peanut oil, and sesame oil.
Journal of AOAC International (2010-07-16)
Lei Bao, Mary W Trucksess, Kevin D White
ABSTRACT
Edible oils are consumed directly, and used as ingredients in food, soaps, and skin products. However, oils such as olive oil, peanut oil, and sesame oil could be contaminated with aflatoxins, which are detrimental to human and animal health. A method using immunoaffinity column cleanup with RPLC separation and fluorescence detection (FLD) for determination of aflatoxins (AF) B1, B2, G1, and G2 in olive oil, peanut oil, and sesame oil was developed and validated. Test samples were extracted with methanol-water (55 + 45, v/v). After shaking and centrifuging, the lower layer was filtered, diluted with water, and filtered through glass microfiber filter paper. The filtrate was then passed through an immunoaffinity column, and the toxins were eluted with methanol. The toxins were then subjected to RPLC/FLD analysis after postcolumn UV photochemical derivatization. The accuracy and repeatability characteristics of the method were determined. Recoveries of AFB1 spiked at levels from 1.0 to 10.0 microg/kg in olive oil, peanut oil, and sesame oil ranged from 82.9 to 98.6%. RSDs ranged from 0.6 to 8.9%. HorRat values were < 0.2 for all of the matrixes tested. Recoveries of AF spiked at levels from 2.0 to 20.0 microg/kg ranged from 87.7 to 102.2%. RSDs ranged from 1.3 to 12.6%. HorRat values were < 0.4 for all of the matrixes tested. LC/MS/MS with multiple-reaction monitoring was used to confirm the identities of aflatoxins in a naturally contaminated peanut oil.
Sigma-Aldrich
Aflatoxin G2
Sigma-Aldrich
Aflatoxin G1, from Aspergillus flavus
Sigma-Aldrich
Aflatoxin B2
Supelco
Aflatoxin G2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Aflatoxin G2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin B2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin G1 solution, 3.78 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin G1 solution, 2 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin B2 solution, 0.5 μg/mL in acetonitrile, analytical standard