Merck
  • Structured triacylglycerol containing behenic and oleic acids suppresses triacylglycerol absorption and prevents obesity in rats.
Structured triacylglycerol containing behenic and oleic acids suppresses triacylglycerol absorption and prevents obesity in rats.
Lipids in health and disease (2010-07-27)
Makiko Kojima, Nobuhiko Tachibana, Takashi Yamahira, Satoshi Seino, Ayako Izumisawa, Nobuo Sagi, Toshiharu Arishima, Mitsutaka Kohno, Kiyoharu Takamatsu, Motohiko Hirotsuka, Ikuo Ikeda
ABSTRACT
Dietary 1(3)-behenoyl-2,3(1)-dioleoyl-rac-glycerol (BOO) has been reported to inhibit pancreatic lipase activity in vitro and suppress postprandial hypertriacylglycerolemia in humans. In the present study, the anti-obesity activities of BOO and its inhibitory effects on lymphatic triacylglycerol (TAG) absorption were investigated in rats. In Experiment 1, rats were fed either BOO or soybean oil (SO) diet for 6 weeks. In the BOO diet, 20% of SO was replaced with an experimental oil rich in BOO. In Experiments 2 and 3, rats cannulated in the thoracic duct were administered an emulsions containing trioleoylglycerol (OOO) or an oil mixture (OOO:BOO, 9:1). Tri[1-14C]oleoylglycerol (14C-OOO) was added to the emulsions administered in Experiment 3. No observable differences were detected in food intake or body weight gain between the BOO and SO groups in Experiment 1. Plasma and liver TAG concentrations and visceral fat weights were significantly lower in the BOO group than in the SO group. The apparent absorption rate of fat was significantly lower in the BOO group than in the SO group. In Experiment 2, the lymphatic recovery of oleic and behenic acids was significantly lower at 5 and 6 h after BOO administration than after OOO administration. In Experiment 3, the lymphatic recovery of 14C-OOO was significantly lower at 5 and 6 h after BOO administration than after OOO administration. These results suggest that BOO prevents deposition of visceral fat and hepatic TAG by lowering and delaying intestinal absorption of TAG.
Sigma-Aldrich
Behenic acid, 99%
Supelco
Behenic acid, analytical standard