- Knocking down the transcript of protein kinase C-lambda modulates hypothalamic glutathione peroxidase, melanocortin receptor and neuropeptide Y gene expression in amphetamine-treated rats.
Knocking down the transcript of protein kinase C-lambda modulates hypothalamic glutathione peroxidase, melanocortin receptor and neuropeptide Y gene expression in amphetamine-treated rats.
It has been reported that neuropeptide Y (NPY) contributes to the behavioral response of amphetamine (AMPH), a psychostimulant. The present study examined whether protein kinase C (PKC)-λ signaling was involved in this action. Moreover, possible roles of glutathione peroxidase (GP) and melanocortin receptor 4 (MC4R) were also examined. Rats were given AMPH daily for 4 days. Hypothalamic NPY, PKCλ, GP and MC4R were determined and compared. Pretreatment with α-methyl-para-tyrosine could block AMPH-induced anorexia, revealing that endogenous catecholamine was involved in regulating AMPH anorexia. PKCλ, GP and MC4R were increased with maximal response on Day 2 during AMPH treatment, which were concomitant with the decreases in NPY. cAMP response element binding protein (CREB) DNA binding activity was increased during AMPH treatment, revealing the involvement of CREB-dependent gene transcription. An interruption of cerebral PKCλ transcript could partly block AMPH-induced anorexia and partly reverse NPY, MC4R and GP mRNA levels to normal. These results suggest that PKCλ participates in regulating AMPH-induced anorexia via a modulation of hypothalamic NPY gene expression and that increases of GP and MC4R may contribute to this modulation. Our results provided molecular evidence for the regulation of AMPH-induced behavioral response.