Merck
A bimetallic iron(III) catalyst for CO2/epoxide coupling.
Chemical communications (Cambridge, England) (2010-09-28)
Antoine Buchard, Michael R Kember, Karl G Sandeman, Charlotte K Williams
ABSTRACT
A novel di-iron(III) catalyst for the copolymerisation of cyclohexene oxide and CO(2) to yield poly(cyclohexene carbonate), under mild conditions, is reported. The catalyst selectivity was completely changed on addition of an ammonium co-catalyst to yield only the cis-isomer of the cyclic carbonate, also under mild conditions. Additionally, the catalyst was active for propylene carbonate and styrene carbonate production at 1 atm pressure.
Sigma-Aldrich
Cyclohexene oxide, 98%