Merck
  • Monitoring membrane binding and insertion of peptides by two-color fluorescent label.
Monitoring membrane binding and insertion of peptides by two-color fluorescent label.
Biochimica et biophysica acta (2010-10-12)
V Y Postupalenko, V V Shvadchak, G Duportail, V G Pivovarenko, A S Klymchenko, Y Mély
ABSTRACT
Herein, we developed an approach for monitoring membrane binding and insertion of peptides using a fluorescent environment-sensitive label of the 3-hydroxyflavone family. For this purpose, we labeled the N-terminus of three synthetic peptides, melittin, magainin 2 and poly-l-lysine capable to interact with lipid membranes. Binding of these peptides to lipid vesicles induced a strong fluorescence increase, which enabled to quantify the peptide-membrane interaction. Moreover, the dual emission of the label in these peptides correlated well with the depth of its insertion measured by the parallax quenching method. Thus, in melittin and magainin 2, which show deep insertion of their N-terminus, the label presented a dual emission corresponding to a low polar environment, while the environment of the poly-l-lysine N-terminus was rather polar, consistent with its location close to the bilayer surface. Using spectral deconvolution to distinguish the non-hydrated label species from the hydrated ones and two photon fluorescence microscopy to determine the probe orientation in giant vesicles, we found that the non-hydrated species were vertically oriented in the bilayer and constituted the best indicators for evaluating the depth of the peptide N-terminus in membranes. Thus, this label constitutes an interesting new tool for monitoring membrane binding and insertion of peptides.
Sigma-Aldrich
Magainin II, ≥97% (HPLC)
Sigma-Aldrich
3-Hydroxyflavone, ≥98%