Merck
  • Ultrastructural alterations induced by Δ(24(25))-sterol methyltransferase inhibitors on Trichomonas vaginalis.
Ultrastructural alterations induced by Δ(24(25))-sterol methyltransferase inhibitors on Trichomonas vaginalis.
FEMS microbiology letters (2010-12-24)
Ivone de Andrade Rosa, Débora Afonso Silva Rocha, Wanderley de Souza, Julio A Urbina, Marlene Benchimol
ABSTRACT
Trichomonas vaginalis is an important human parasite that causes trichomoniasis, a cosmopolitan sexually transmitted disease. Currently, the treatment of choice for T. vaginalis infections is metronidazole. The increase in metronidazole-resistant parasites and undesirable side effects of this drug make the search for alternative chemotherapeutic approaches a priority for the management of trichomoniasis. Here, the antiproliferative and ultrastructural effects of sterol biosynthesis inhibitors against T. vaginalis were investigated. It was found that 22,26-azasterol (5 μM) and 24(R,S),25-epiminolanosterol (10 μM), known inhibitors of Δ(24(25))-sterol methyltransferase, exhibited antiproliferative effects on T. vaginalis trophozoites cultured in vitro. Morphological analyses showed that azasterols induced changes in the ultrastructure of T. vaginalis. The most significant alterations were (1) membrane blebbing and disruption, (2) wrinkled cells and (3) the formation of cell clusters. In addition, autophagic vacuoles, Golgi duplication arrest, an abnormal Golgi enlargement and damaged hydrogenosomes were also observed. Nonspecific cytotoxicity assays using the cultured mammalian cell lines Madin-Darby canine kidney cells showed no effect of the azasterols on the viability and proliferation of these cells at a concentration that significantly inhibited the proliferation of T. vaginalis, indicating a selective antiparasitic action. Taken together, these results suggest that azasterols could be important compounds in the development of novel chemotherapeutic approaches against T. vaginalis.
Sigma-Aldrich
5β-Cholestan-3α-ol, ≥95%
Sigma-Aldrich
5α-Cholestan-3β-ol, ≥95%
Sigma-Aldrich
Coprostan-3-ol, ≥98%