Merck
  • Palladium-catalyzed selective acyloxylation using sodium perborate as oxidant.
Palladium-catalyzed selective acyloxylation using sodium perborate as oxidant.
The Journal of organic chemistry (2011-01-22)
Lukasz T Pilarski, Pär G Janson, Kálmán J Szabó
ABSTRACT
Sodium perborate (SPB), a principal component of washing powders, was employed as an inexpensive and eco-friendly oxidant in the palladium-catalyzed C-H acyloxylation of alkenes in excellent regio- and stereochemistry. The reactions used anhydrides as acyloxy sources. The method applies to both terminal and internal alkenes, and even benzylic C-H oxidation.
Sigma-Aldrich
Sodium perborate monohydrate, powder
Sigma-Aldrich
Sodium perborate tetrahydrate, 9-11% available Oxygen
Sigma-Aldrich
Sodium perborate tetrahydrate, purum p.a., 96% (RT)