Merck
  • Synthesis of sterically congested 1,3,4-oxadiazole derivatives from aromatic carboxylic acids, N,N-dicyclohexylcarbodiimide and (N-isocyanimino)triphenylphosphorane.
Synthesis of sterically congested 1,3,4-oxadiazole derivatives from aromatic carboxylic acids, N,N-dicyclohexylcarbodiimide and (N-isocyanimino)triphenylphosphorane.
Molecular diversity (2011-03-23)
Fatemeh Zeinali Nasrabadi, Ali Ramazani, Yavar Ahmadi
ABSTRACT
Reactions of (N-isocyanimino)triphenylphosphorane with N,N-dicyclohexylcarbodiimide in the presence of aromatic (or heteroaromatic) carboxylic acids proceed smoothly at room temperature and in neutral conditions to afford sterically congested 1,3,4-oxadiazole derivatives in high yields. The reaction proceeds smoothly and cleanly under mild conditions, and no side reactions were observed.
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
DCC, 99%