Merck
  • Determination of specific neuropeptides modulation time course in a rat model of osteoarthritis pain by liquid chromatography ion trap mass spectrometry.
Determination of specific neuropeptides modulation time course in a rat model of osteoarthritis pain by liquid chromatography ion trap mass spectrometry.
Neuropeptides (2011-08-23)
Catherine E Ferland, Floriane Pailleux, Pascal Vachon, Francis Beaudry
ABSTRACT
Animal models are useful to evaluate pharmacological therapies to alleviate joint pain. The present study characterized central neuropeptides modulation in the monoiodoacetate (MIA) rat model. Animals receiving a single 3mg MIA injection were euthanized at 3, 7, 14, 21 and 28 days post injection. Spinal cords were analyzed by liquid chromatography ion trap mass spectrometry. Up-regulations of the calcitonin gene-related peptide and substance P were observed starting on days 7 and 28 respectively, whereas big dynorphin(₁₋₃₂) content decreased significantly on day 14 in comparison to control animals (P<0.05). Preclinical drug evaluations using this model should be conducted between 7 and 21 days post injection when the lesions resemble most to human osteoarthritis.
Sigma-Aldrich
Iodoacetic acid, ≥98.0% (T)