Merck
A π-stacked phenylacetylene dimer.
Physical chemistry chemical physics : PCCP (2011-08-23)
Surajit Maity, G Naresh Patwari, Robert Sedlak, Pavel Hobza
ABSTRACT
The structure of the phenylacetylene-dimer has been elucidated using IR-UV double resonance spectroscopy in combination with high level ab initio calculations at the CCSD(T)/CBS level. The IR spectra in the acetylenic and the aromatic C-H stretching regions indicate that the two phenylacetylene moieties are in identical environments and very similar to the phenylacetylene monomer. Calculated stabilization energies and the free energies at the CCSD(T)/CBS level favor the formation of an anti-parallel π-stacked structure. The DFT-SAPT energy decomposition analysis points out that the anti-parallel π-stacked structure maximizes electrostatic as well as the dispersion components of energy. The observed IR spectra are consistent with the anti-parallel π-stacked structure.
Sigma-Aldrich
Phenylacetylene, 98%