Merck
  • Prenatal developmental toxicity studies on di-n-heptyl and di-n-octyl phthalates in Sprague-Dawley rats.
Prenatal developmental toxicity studies on di-n-heptyl and di-n-octyl phthalates in Sprague-Dawley rats.
Reproductive toxicology (Elmsford, N.Y.) (2011-09-13)
Anne-Marie Saillenfait, Alain-Claude Roudot, Frédéric Gallissot, Jean-Philippe Sabaté
ABSTRACT
This study evaluates the developmental toxicity of two dialkyl phthalate esters, di-n-heptyl phthalate (DHPP) and di-n-octyl (DnOP) phthalate, which have straight-alkyl side chains of seven and eight carbons, respectively. Sprague-Dawley rats were administered 0, 0.25, 0.50, or 1g/kg/day of DHPP or DnOP, by gavage, on gestation days 6-20. DHPP and DnOP had no adverse effect on maternal feed consumption and body weight gain, or on the incidence of post-implantation loss and fetal body weight. There was no increase in the incidence of fetal malformations or external and visceral variations, whatever treatment. A significant increase in rudimentary lumbar ribs was observed at all doses of DHPP and DnOP. The anogenital distance of the male fetuses was significantly decreased at the highest dose of DHPP. This parameter was not affected by DnOP. Thus, the lowest-observed-adverse-effect level (LOAEL) for developmental toxicity was 0.25 g/kg/day for DHPP and DnOP.
Product Description
Supelco
Di-n-octyl phthalate, analytical standard
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Sigma-Aldrich
Diheptyl phthalate, 97%
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland