Merck
Antibacterial nanofiber materials activated by light.
Journal of biomedical materials research. Part A (2011-10-06)
Soňa Jesenská, Lukáš Plíštil, Pavel Kubát, Kamil Lang, Libuše Brožová, Stěpán Popelka, Lórant Szatmáry, Jiří Mosinger
ABSTRACT
Electrospun polymeric nanofiber materials doped with 5,10,15,20-tetraphenylporphyrin (TPP) photosensitizer were prepared from four different polymers and were characterized with microscopic methods, steady-state, and time-resolved fluorescence and absorption spectroscopy. The polymers used included polyurethane Larithane™ (PUR), polystyrene (PS), polycaprolactone (PCL), and polyamide 6 (PA6). The antibacterial activity of all nanofiber materials against E. coli was activated by visible light and it was dependent on oxygen permeability/diffusion coefficients and the diameter of the polymeric nanofibers. This activity is based on oxidation ability of singlet oxygen O₂(¹Δ(g)) that is generated upon irradiation. All tested nanofiber materials exhibited prolonged antibacterial properties, even in the dark after long-duration irradiation. The post-irradiation effect was explained by the photogeneration of H₂O₂, which provided the material with long-lasting antibacterial properties.
Sigma-Aldrich
Nylon 6, pellets