Merck
  • Synthesis and histamine H(3) and H(4) receptor activity of conformationally restricted cyanoguanidines related to UR-PI376.
Synthesis and histamine H(3) and H(4) receptor activity of conformationally restricted cyanoguanidines related to UR-PI376.
Archiv der Pharmazie (2011-10-12)
Roland Geyer, Armin Buschauer
ABSTRACT
Recently, we identified highly potent agonists of the human histamine H(4) receptor (hH(4) R) among a series of imidazolylbutylcyanoguanidines. Aiming at improved selectivity for the hH(4) R relative to the H(3) receptor (hH(3) R), the flexible tetramethylene linker connecting imidazole ring and cyanoguanidine group was replaced by conformationally restricted carbocycles. Introduction of a para- or a meta-phenylene spacer yielded only very weakly active compounds at both hH(3) R and hH(4) R (investigated in [(35) S]GTPγS binding assays using Sf9 insect cell membranes expressing hH(x) R subtypes). By contrast, the incorporation of a more flexible cyclohexane-1,4-diyl linker resulted in EC(50) or K(B) values ≥110 nM at hH(4) R and hH(3) R. Quality of action, potency and receptor subtype selectivity of the investigated compounds depend on the stereochemistry: Cis-configured diastereomers prefer the hH(4) R and are partial agonists, whereas trans-isomers are antagonists at the hH(4) R. At the hH(3) R the trans-diastereomers are superior to the cis-isomers by a factor of 10. The results on imidazolylcycloalkylcyanoguanidines suggest that variation of ring size and optimization of the stereochemistry may be useful to increase the potency and selectivity of hH(4) R agonists relative to the hH(3) R.
Sigma-Aldrich
Dicyandiamide, 99%