Merck
  • A molecular chalice with hydrophobic walls and a hydrophilic rim: self-assembly and complexation properties.
A molecular chalice with hydrophobic walls and a hydrophilic rim: self-assembly and complexation properties.
Chemical communications (Cambridge, England) (2011-11-03)
Marc Vidal, Vanessa Kairouz, Christine DeWolf, Andreea R Schmitzer
ABSTRACT
We describe the synthesis of a diphenylglycoluril/dibenzo-crown-6 molecular chalice, the self-assembly at the air/water interface and its complexation properties in solution and at the water/chloroform interface.
Sigma-Aldrich
Dibenzo-18-crown-6, 98%