Merck
  • Lanthanum chloride bidirectionally influences calcification in bovine vascular smooth muscle cells.
Lanthanum chloride bidirectionally influences calcification in bovine vascular smooth muscle cells.
Journal of cellular biochemistry (2012-01-04)
Wen-Hua Zhao, Bao-Di Gou, Tian-Lan Zhang, Kui Wang
ABSTRACT
Vascular calcification (VC) is frequent prevalence in patients with chronic kidney disease (CKD) and atherosclerosis. Lanthanum carbonate is used as an orally administered phosphate-binding agent to reduce the gastrointestinal absorption of phosphate and ameliorate VC in advanced CKD. In this study, we used bovine vascular smooth muscle cells as a model VC in vitro and studied the effects of lanthanum chloride on calcium deposition. Exposure of cells to LaCl(3) at the concentration of 0.1 µM suppressed the β-glycerophosphate-induced alkaline phosphatase activity and calcium deposition. Furthermore, LaCl(3) upregulated the β-glycerophosphate-suppressed expression of calcium-sensing receptor. In contrast to the inhibitory effect of LaCl(3) on calcium deposition, higher level lanthanum (50 µM) was found to promote immediately precipitation of calcium phosphate in cell culture medium. At this concentration, LaCl(3) was found to induce cell apoptosis which involves caspases-9 and -3. These data indicate that the promotory effect of LaCl(3) on calcium deposition is likely mediated by induction of apoptosis. Our in vitro findings do suggest that, in the context of raised lanthanum, greater attention should be paid to potential toxic effects associated to the use of lanthanide-based drugs.
Sigma-Aldrich
Glycerol phosphate calcium salt, >98% (TLC)
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, ACS reagent
Sigma-Aldrich
Lanthanum(III) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis