Merck
  • Seed germination responses to varying environmental conditions and provenances in Crucianella maritima L., a threatened coastal species.
Seed germination responses to varying environmental conditions and provenances in Crucianella maritima L., a threatened coastal species.
Comptes rendus biologies (2012-01-10)
Silvia Del Vecchio, Efisio Mattana, Alicia T R Acosta, Gianluigi Bacchetta
ABSTRACT
Seed germination (effects of light, temperature, NaCl and KNO(3)) of the coastal endangered species Crucianella maritima was investigated by testing seeds from three different populations. Data were analyzed by means of Generalized Linear Mixed Model (GLMM). The principal results showed that germination of C. maritima seeds was characterized by photoinhibition, absence of primary dormancy and salt-induced secondary dormancy, with no need for high nutrient availability (KNO(3)). Intraspecific differences in germination pattern emerged, apparently due to a different seed mass. These results show important germination traits of C. maritima which should be taken into account in possible reintroduction attempts aimed at restoring threatened populations of this species.
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)