Merck
  • Olfactory responses to explosives associated odorants are enhanced by zinc nanoparticles.
Olfactory responses to explosives associated odorants are enhanced by zinc nanoparticles.
Talanta (2012-01-24)
Christopher H Moore, Oleg Pustovyy, John C Dennis, Timothy Moore, Edward E Morrison, Vitaly J Vodyanoy
ABSTRACT
Many odorants related to manufactured explosives have low volatilities and are barely detectable as odors. We previously reported that zinc metal nanoparticles increased rat olfactory epithelium responses, measured by electroolfactogram (EOG), to several odorants. Here, we report that nanomolar concentrations of zinc metal nanoparticles strongly enhanced olfactory responses to the explosives related odorants cyclohexanone, methyl benzoate, acetophenone, and eugenol. Rat olfactory epithelium was exposed to metal nanoparticles and odorant responses were quantified by EOG. Zinc nanoparticles added to explosive odorants strongly increased the odorant response in a dose-dependent manner. The enzymatic breakdown of the second messenger cyclic adenosine monophosphate (cAMP) was prevented by adding the membrane-permeable phosphodiesterase inhibitor 3-isobutyl-1-methylxanthine (IBMX). This caused the olfactory cilia cAMP concentration to increase and generated EOG signals. The EOG responses generated by IBMX were not enhanced by zinc nanoparticles. Based on these observations, we conclude that zinc nanoparticles act at the receptor site and are involved in the initial events of olfaction. Our results suggest that zinc metal nanoparticles can be used to facilitate a canine detection of explosive odorants.
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Supelco
Cyclohexanone, Selectophore, ≥99.5%
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Methyl benzoate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Supelco
Methyl benzoate, analytical standard
Sigma-Aldrich
Methyl benzoate, ≥98%, FCC, FG
Sigma-Aldrich
Cyclohexanone, 99.8%
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Methyl benzoate, 99%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)