Merck
  • Synthesis, characterization and chemoprotective activity of polyoxovanadates against DNA alkylation.
Synthesis, characterization and chemoprotective activity of polyoxovanadates against DNA alkylation.
Journal of inorganic biochemistry (2012-01-24)
Giovana G Nunes, Ana C Bonatto, Carla G de Albuquerque, Andersson Barison, Ronny R Ribeiro, Davi F Back, André Vitor C Andrade, Eduardo L de Sá, Fábio de O Pedrosa, Jaísa F Soares, Emanuel M de Souza
ABSTRACT
The alkylation of pUC19 plasmid DNA has been employed as a model reaction for the first studies on chemoprotective action by a mixed-valence (+IV/+V) polyoxovanadate. A new, non-hydrothermal route for the high yield preparation of the test compound is described. The deep green, microcrystalline solid A was isolated after a three-day reaction in water at 80°C and 1 atm, while the reaction at 100°C gave green crystals of B. Both solids were structurally characterized by X-ray diffractometry and FTIR, EPR, NMR and Raman spectroscopies. Product A was identified as (NH(4))(2)V(3)O(8), while B corresponds to the spherical polyoxoanion [V(15)O(36)(Cl)](6-), isolated as the NMe(4)(+) salt. The lack of solubility of A in water and buffers prevented its use in DNA interaction studies, which were then carried out with B. Complex B was also tested for its ability to react with DNA alkylating agents by incubation with diethylsulphate (DES) and dimethylsulphate (DMS) in both the absence and presence of pUC19. For DMS, the best results were obtained with 10 mM of B (48% protection); with DES, this percentage increased to 70%. The direct reaction of B with increasing amounts of DMS in both buffered (PIPES 50 mM) and non-buffered aqueous solutions revealed the sequential formation of several vanadium(IV), vanadium(V) and mixed-valence aggregates of different nuclearities, whose relevance to the DNA-protecting activity is discussed.
Sigma-Aldrich
Diethyl sulfate, 98%