Merck
  • An artificial photosensitizer drug network for mitochondria-selective photodynamic therapy.
An artificial photosensitizer drug network for mitochondria-selective photodynamic therapy.
Chemical communications (Cambridge, England) (2012-01-31)
So Young Park, Kyung Taek Oh, Young Taik Oh, Nam Muk Oh, Yu Seok Youn, Eun Seong Lee
ABSTRACT
We report a new class of photosensitizer drug networks that can home into mitochondria and provide geospatial phototoxicity for tumors. We take advantage of the cleavable chemical network between the photosensitizer drug and poly(ethylene glycol), and find a significant increase in the efficiency of the multimeric drug network in mitochondria uptake and tumor inhibition.
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride