Merck
  • Long-term protective effect of surface sealants against erosive wear by intrinsic and extrinsic acids.
Long-term protective effect of surface sealants against erosive wear by intrinsic and extrinsic acids.
Journal of dentistry (2012-02-14)
Florian J Wegehaupt, Tobias T Tauböck, Beatrice Sener, Thomas Attin
ABSTRACT
To test sealants to prevent erosive tooth wear caused by extrinsic and intrinsic acids under long-term exposition. 144 bovine enamel samples were randomly allocated to twelve groups (1-12). Samples of groups 1, 5 and 9 remained unsealed (positive controls), 2, 6 and 10 were sealed with Silicon Seal Nano Mix and 3, 7 and 11 with Seal&Protect. Groups 4, 8 and 12 were sealed with flowable composite (negative controls). Groups 1-4 were immersed in artificial saliva, 5-8 in hydrochloric acid and groups 9-12 in citric acid for 28 days, respectively. After 1, 2, 4, 7, 11, 14, 21 and 28 days, solutions were renewed and enamel wear was quantified by assignation of (32)P in the solutions. In all immersion solutions, lowest mineral loss was observed for the negative controls whilst highest loss was observed for unsealed positive controls. In artificial saliva and citric acid, the loss from samples sealed with Seal&Protect was not significantly different compared with negative controls whilst loss in groups sealed with Silicon Seal Nano Mix was significantly higher. In hydrochloric acid, loss from samples sealed with Seal&Protect was not different compared with that of negative controls up to 4 days. Except day 1, the mineral loss in the Seal&Protect group was significantly lower compared with that of the Silicon Seal Nano Mix group. The tested resin based surface sealant is able to significantly reduce the erosive demineralisation of enamel caused by hydrochloric and citric acid even under long-term exposition.
Sigma-Aldrich
Calcium fluoride, random crystals, 99.99% trace metals basis
Sigma-Aldrich
Calcium fluoride, powder, 99.99% trace metals basis