Merck
Syntheses of (-)-pelletierine and (-)-homopipecolic acid.
Organic & biomolecular chemistry (2012-02-22)
Wen-Hua Chiou, Guei-Tang Chen, Chien-Lun Kao, Yu-Kai Gao
ABSTRACT
Enantiomeric syntheses of (-)-homopipecolic acid and (-)-pelletierine have been achieved by chiral resolution of tropanol followed by Baeyer-Villiger oxidation. The methodology provides a practical route for the synthesis of optically pure piperidines.
Sigma-Aldrich
Pipecolinic acid, 98%
Sigma-Aldrich
L-Pipecolic acid, 99% (titration)