Merck
  • NIPA1 polyalanine repeat expansions are associated with amyotrophic lateral sclerosis.
NIPA1 polyalanine repeat expansions are associated with amyotrophic lateral sclerosis.
Human molecular genetics (2012-03-02)
Hylke M Blauw, Wouter van Rheenen, Max Koppers, Philip Van Damme, Stefan Waibel, Robin Lemmens, Paul W J van Vught, Thomas Meyer, Claudia Schulte, Thomas Gasser, Edwin Cuppen, R Jeroen Pasterkamp, Wim Robberecht, Albert C Ludolph, Jan H Veldink, Leonard H van den Berg
ABSTRACT
Mutations in NIPA1 cause Hereditary Spastic Paraplegia type 6, a neurodegenerative disease characterized by an (upper) motor neuron phenotype. Deletions of NIPA1 have been associated with a higher susceptibility to amyotrophic lateral sclerosis (ALS). The exact role of genetic variation in NIPA1 in ALS susceptibility and disease course is, however, not known. We sequenced the entire coding sequence of NIPA1 and genotyped a polyalanine repeat located in the first exon of NIPA1. A total of 2292 ALS patients and 2777 controls from three independent European populations were included. We identified two sequence variants that have a potentially damaging effect on NIPA1 protein function. Both variants were identified in ALS patients; no damaging variants were found in controls. Secondly, we found a significant effect of 'long' polyalanine repeat alleles on disease susceptibility: odds ratio = 1.71, P = 1.6 × 10(-4). Our analyses also revealed a significant effect of 'long' alleles on patient survival [hazard ratio (HR) = 1.60, P = 4.2 × 10(-4)] and on the age at onset of symptoms (HR = 1.37, P = 4.6 × 10(-3)). In patients carrying 'long' alleles, median survival was 3 months shorter than patients with 'normal' genotypes and onset of symptoms occurred 3.6 years earlier. Our data show that NIPA1 polyalanine repeat expansions are a common risk factor for ALS and modulate disease course.
Sigma-Aldrich
Poly-DL-alanine, mol wt 1,000-5,000