Merck
  • An anatomically based approach to intralesional corticosteroid injection for eyelid capillary hemangiomas.
An anatomically based approach to intralesional corticosteroid injection for eyelid capillary hemangiomas.
Ophthalmic surgery, lasers & imaging : the official journal of the International Society for Imaging in the Eye (2012-03-22)
David B Samimi, Chrisfouad R Alabiad, David T Tse
ABSTRACT
To describe a corticosteroid injection technique for eyelid capillary hemangiomas that minimizes the risk of a central retinal artery occlusion and is based on anatomic, physiologic, and pharmacologic rationales. In this retrospective, non-comparative, interventional case series, the medical records of 50 eyes of 50 patients over a 10-year period were reviewed for retinal complications associated with the described injection technique. Ophthalmoscopy showed no retinal complications in the 50 eyes treated with this injection technique. The risk of central retinal artery occlusion from retrograde embolization can be minimized by using an anatomically based injection technique that prevents canalization of an artery and avoids injection pressures exceeding the mean systemic arterial pressure.
Sigma-Aldrich
Betamethasone 21-phosphate disodium, ≥97%
Betamethasone sodium phosphate, European Pharmacopoeia (EP) Reference Standard