Merck
  • Recombinant expression, in vitro refolding, and biophysical characterization of the N-terminal domain of T1R3 taste receptor.
Recombinant expression, in vitro refolding, and biophysical characterization of the N-terminal domain of T1R3 taste receptor.
Protein expression and purification (2012-03-28)
Elodie Maîtrepierre, Maud Sigoillot, Laurence Le Pessot, Loïc Briand
ABSTRACT
The sweet taste receptor is a heterodimeric receptor composed of the T1R2 and T1R3 subunits, while T1R1 and T1R3 assemble to form the umami taste receptor. T1R receptors belong to the family of class C G-protein coupled receptors (GPCRs). In addition to a transmembrane heptahelical domain, class C GPCRs have a large extracellular N-terminal domain (NTD), which is the primary ligand-binding site. The T1R2 and T1R1 subunits have been shown to be responsible for ligand binding, via their NTDs. However, little is known about the contribution of T1R3-NTD to receptor functions. To enable biophysical characterization, we overexpressed the human NTD of T1R3 (hT1R3-NTD) using Escherichia coli in the form of inclusion bodies. Using a fractional factorial screen coupled to a functional assay, conditions were determined for the refolding of hT1R3-NTD. Far-UV circular dichroism spectroscopic studies revealed that hT1R3-NTD was well refolded. Using size-exclusion chromatography, we found that the refolded protein behaves as a dimer. Ligand binding quantified by tryptophan fluorescence quenching and microcalorimetry showed that hT1R3-NTD is functional and capable of binding sucralose with an affinity in the millimolar range. This study also provides a strategy to produce functional hT1R3-NTD by heterologous expression in E. coli; this is a prerequisite for structural determination and functional analysis of ligand-binding regions of other class C GPCRs.
Product Description
Supelco
Sucralose, analytical standard
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)