Merck
  • A novel flow-injection analysis system for evaluation of antioxidants by using sodium dichloroisocyanurate as a source of hypochlorite anion.
A novel flow-injection analysis system for evaluation of antioxidants by using sodium dichloroisocyanurate as a source of hypochlorite anion.
Drug discoveries & therapeutics (2012-03-31)
H Ichiba, K Hanami, K Yagasaki, M Tanaka, H Ito, T Fukushima
ABSTRACT
A flow injection analysis (FIA) system for evaluation of the antioxidant activity of a compound capable of scavenging a hypochlorite anion (OCl⁻), one of the reactive oxygen species (ROS), was developed. Aminophenyl fluorescein (APF), a fluorescence indicator of ROS, was mixed manually with the test compounds and the mixed solution was injected into the FIA system. The injected solution was reacted in-line with OCl⁻, that was produced by using sodium dichloroisocyanurate in the presence of 0.1 M CH3CO2Na in H2O. The fluorescence intensity of fluorescein generated from non-fluorescent APF was significantly attenuated in compounds that had a scavenging effect on OCl⁻. The precision obtained by the FIA system was satisfactory (relative standard deviation < 5.0%) and a rapid assay within 0.5 min per sample was achieved. The proposed FIA system was used to demonstrate that reduced glutathione, dithiothreitol, and 3-methyl-1-phenyl-5-pyrazolone (edaravone) showed a significant scavenging effect on OCl⁻. Therefore, the proposed FIA system can be used as a screening assay for OCl⁻-scavenging compounds.
Sigma-Aldrich
Sodium dichloroisocyanurate, 96%
Sigma-Aldrich
Sodium dichloroisocyanurate dihydrate, 98.0% (AT)