Merck
Tyrosine-like condensed derivatives as tyrosinase inhibitors.
The Journal of pharmacy and pharmacology (2012-04-05)
Maria João Matos, Lourdes Santana, Eugenio Uriarte, Silvia Serra, Marcella Corda, Maria Benedetta Fadda, Benedetta Era, Antonella Fais
ABSTRACT
We report the pharmacological evaluation of a new series of 3-aminocoumarins differently substituted with hydroxyl groups, which have been synthesized because they include in their structures the tyrosine fragment (tyrosine-like compounds), with the aim of discovering structural features necessary for tyrosinase inhibitory activity. The synthesized compounds 4 and 7-9 were evaluated in vitro as mushroom tyrosinase inhibitors. Two of the described compounds showed lower IC50 (concentration giving 50% inhibition of tyrosinase activity) than umbelliferone, used as a reference compound. Compound 7 (IC50=53µm) was the best tyrosinase inhibitor of this small series, having an IC50 value 10-fold lower than umbelliferone. Compound 7 (3-amino-7-hydroxycoumarin) had amino and hydroxyl groups precisely mimicking the same positions that both groups occupy on the tyrosine molecule.
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Supelco
Umbelliferone, analytical standard