Merck
CuBr2--a new multiferroic material with high critical temperature.
Advanced materials (Deerfield Beach, Fla.) (2012-04-11)
Li Zhao, Tsu-Lien Hung, Ching-Chien Li, Yang-Yuan Chen, Maw-Kuen Wu, Reinhard K Kremer, Michael G Banks, Arndt Simon, Myung-Hwan Whangbo, Changhoon Lee, Jun Sung Kim, Ingyu Kim, Kee Hoon Kim
ABSTRACT
A new multiferroic material, CuBr(2) , is reported for the first time. CuBr(2) has not only a high transition temperature (close to liquid nitrogen temperature) but also low dielectric loss and strong magnetoelectric coupling. These findings reveal the importance of anion effects, in the search for the high temperature multiferroics materials among these low-dimensional spin systems.
Sigma-Aldrich
Copper(II) bromide, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) bromide, 99%