Merck
  • [Analyze on volatile compounds of Antrodia camphorata using HS-SPME-GC-MS].
[Analyze on volatile compounds of Antrodia camphorata using HS-SPME-GC-MS].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-04-18)
Zhe He, Zhen-Ming Lu, Hong-Yu Xu, Jing-Song Shi, Zheng-Hong Xu
ABSTRACT
To analyze the volatile compounds of Antrodia camphorata in solid-state and submerged cultures. A headspace solid-phase microextraction (HS-SPME) coupled with gas chromatography-mass spectrometry(GC-MS) were used to evaluate the profile of the volatile compounds. 49 volatile compounds were identified in A. camphorata mycelia in submerged culture, while 43 volatile compounds were identified in mycelia in solid-state culture. 1-octen-3-ol, 3-octanone, 1-octen-3-ylacetate, acetic acid octyl ester and ethanol were the main volatile compounds in A. camphorata mycelia in submerged culture, while 1-octen-3-ol, 3-octanone, 3-methyl-butyraldenhyde, gamma-podecalactone and methyl 2-furozte were the most potent key volatile compounds in mycelia in solid-state culture. The volatile compounds in the mycelia of A. camphorata in solid-state and submerged cultures are similar but their relative contents are different.
Sigma-Aldrich
3-Octanone, ≥98%, FG
Sigma-Aldrich
3-Octanone, ≥98%
Supelco
3-Octanone, analytical standard
Sigma-Aldrich
Octyl acetate, ≥98%, FCC, FG
Sigma-Aldrich
Octyl acetate, ≥99%
Sigma-Aldrich
Octyl acetate, natural, ≥98%, FCC, FG
Sigma-Aldrich
1-Octen-3-ol, 98%
Sigma-Aldrich
1-Octen-3-ol, ≥98%, FCC, FG