Merck
  • [Analyze on chemical compositions of Dalbergia odorifera essential oils extracted by CO2-supercritical-fluid-extraction and steam distillation extraction].
[Analyze on chemical compositions of Dalbergia odorifera essential oils extracted by CO2-supercritical-fluid-extraction and steam distillation extraction].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-04-18)
Wei-Feng Song, Mei-Jin Liao, Shu-Yuan Luo
ABSTRACT
To analyze the chemical compositions of Dalbergia odorifera essential oils extacted by CO2-supercritical-fluid-extraction (SFE-CO2) and steam distillation extraction (SD). The essential oils of Dalbergia odorifera were extracted by steam distillation extraction and SFE-CO2. The chemical components were separated and analyzed by gas chromatography-mass spectrometry. 12 compounds were identified in SFE sample. The major components from essential oils were 2-propenoic acid-3(4-methoxyphenyl)-ethyl ester (14.53%), nerolidol (14.95%), ageratochromene (1.33%). 9 compounds were identified in SD sample. The major components from essential oils were nerolidol (26.61%), cedrol (1.65%). The SFE method is better than the SD method in reliability stability and reproducibility, and suitable for essential oils extraction of Dalbergia odorifera.
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans
Sigma-Aldrich
(+)-Cedrol, ≥99.0% (sum of enantiomers, GC)
Supelco
trans-Nerolidol, analytical standard