Merck
  • Cu(II)-catalyzed asymmetric hydrosilylation of diaryl- and aryl heteroaryl ketones: application in the enantioselective synthesis of orphenadrine and neobenodine.
Cu(II)-catalyzed asymmetric hydrosilylation of diaryl- and aryl heteroaryl ketones: application in the enantioselective synthesis of orphenadrine and neobenodine.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-05-11)
Yao-Zong Sui, Xi-Chang Zhang, Jun-Wen Wu, Shijun Li, Ji-Ning Zhou, Min Li, Wenjun Fang, Albert S C Chan, Jing Wu
ABSTRACT
With certain amounts of sodium tert-butoxide and tert-butanol as additives, catalytic amounts of an inexpensive and easy-to-handle copper source Cu(OAc)(2)⋅H(2)O, a commercially available and air-stable non-racemic dipyridylphosphine ligand, as well as the stoichiometric desirable hydride donor polymethylhydrosiloxane (PMHS), formed a versatile in situ catalyst system for the enantioselective reduction of a broad spectrum of prochiral diaryl and aryl heteroarylketones in air, in high yields and with good to excellent enantioselectivities (up to 96 %). In particular, the practical viability of this process was evinced by its successful applications in the asymmetric synthesis of optically enriched potent antihistaminic drugs orphenadrine and neobenodine.
Product Description
Supelco
Orphenadrine citrate salt, analytical standard
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
Orphenadrine hydrochloride, ≥98.0% (AT)
Orphenadrine for peak identification, European Pharmacopoeia (EP) Reference Standard