Merck
RNA in situ hybridization in Arabidopsis.
Methods in molecular biology (Clifton, N.J.) (2012-05-17)
Miin-Feng Wu, Doris Wagner
ABSTRACT
RNA in situ hybridization using digoxigenin-labeled riboprobes on tissue sections is a powerful technique for revealing microscopic spatial gene expression. Here, we describe an in situ hybridization method commonly practiced in Arabidopsis research labs. The highly stringent hybridization condition eliminates the usage of Ribonlucease A and gives highly specific signals. This also allows the use of longer probes which enhance signal strength without cross hybridization to closely related genes. In addition, using spin columns in template and riboprobe purification greatly reduces background signals.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Supelco
Digoxigenin, analytical standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard