Merck
  • Acute toxicity of sodium metabisulphite in larvae and post-larvae of the land crab, Cardisoma guanhumi.
Acute toxicity of sodium metabisulphite in larvae and post-larvae of the land crab, Cardisoma guanhumi.
Bulletin of environmental contamination and toxicology (2012-05-31)
Orlando B S Galli, Rodrigo Y Fujimoto, Fernando A Abrunhosa
ABSTRACT
Sodium metabisulphite (SMB) is used in marine shrimp aquaculture to prevent the occurrence of black spot. The release SMB into the estuarine environment from shrimp farm pond effluents has been reported. This study evaluated the susceptibility of larvae and post-larvae of land crab, Cardisoma guanhumi to this salt. A decrease in dissolved oxygen and pH occurred with increasing concentration of SMB and exposure time. LC(50) values after 48 h of exposure were 34 ± 1.1 mg/L, 31.1 ± 1.9 mg/L, and 30.6 ± 0.5 mg/L for I zoea larvae, megalopa larvae and stage I juveniles, respectively.
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%
Supelco
Sodium metabisulfite, analytical standard