Merck
[Halitosis in oral diseases].
Stomatologiia (2012-06-09)
D N Bakhmutov, O I Kharchenko, O O Ianushevich
ABSTRACT
The presence of hydrogen sulfide, methilmercaptane, dimethilsulfide, putrescine, cadaverine in exhaled air was evaluated in the study. The concentrations of the named substances were defined in various oral diseases. The received results are of interest for differential diagnostics of halitosis causes.
Sigma-Aldrich
Sodium methanethiolate, technical, ≥90% (RT)
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Sodium thiomethoxide, 95%
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Supelco
Dimethyl sulfide, analytical standard