Merck
  • An organocatalytic cascade approach toward polysubstituted quinolines and chiral 1,4-dihydroquinolines-unanticipated effect of N-protecting groups.
An organocatalytic cascade approach toward polysubstituted quinolines and chiral 1,4-dihydroquinolines-unanticipated effect of N-protecting groups.
Angewandte Chemie (International ed. in English) (2012-06-15)
Xinshuai Zhang, Xixi Song, Hao Li, Shilei Zhang, Xiaobei Chen, Xinhong Yu, Wei Wang
ABSTRACT
A matter of protection: The outcome of a divergent organocatalytic aza-Michael/aldol cascade process toward quinolines and 1,4-dihydroquinolines depends on the choice of the N-protecting group (see scheme; TEA = triethylamine, TMS = trimethylsilyl). Use of an electron-donating sulfonyl group results in an unanticipated aza-Michael/aldol/aromatization cascade to give polysubstituted quinolines (right). In contrast, chiral 1,4-dihydroquinolines are obtained with an electron-withdrawing sulfonyl group (left).
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)