Merck
  • Lack of CYP1A responsiveness in species inhabiting chronically contaminated habitats: two varieties of resistance?
Lack of CYP1A responsiveness in species inhabiting chronically contaminated habitats: two varieties of resistance?
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2012-07-11)
Ben F Brammell, David J Price, Wesley J Birge, Adria A Elskus
ABSTRACT
Organisms chronically exposed to organic pollutants such as polychlorinated biphenyls (PCBs) can develop resistance to these chemicals, a condition associated with reduced inducibility of the biomarker enzyme cytochrome P450 1A (CYP1A). This study addresses the CYP1A response of members of the families Ictaluridae and Centrarchidae, two fish families found throughout much of the United States. We measured CYP1A expression, PCB body burdens, and conducted CYP1A challenge experiments in species from these families residing in the Town Branch/Mud River system (Logan County, KY, USA), a stream system historically contaminated with high levels of PCBs. Despite PCB concentrations in muscle tissue typically associated with elevated CYP1A (16.7 to 75.2μgPCB/g wet edible flesh), resident fish in the contaminated Town Branch/Mud River sites (yellow bullhead [Ameiurus natalis], green sunfish [Lepomis cyanellus], and spotted bass [Micropterus punctulatus]) had hepatic CYP1A activity levels similar to, rather than higher than, those in reference fish, suggesting reduced sensitivity to CYP1A induction. Lack of CYP1A expression following direct contaminant exposure has often been associated with resistance to those contaminants. To determine if CYP1A in resident populations was resistant to induction by PCBs, we exposed resident fish to a single, intraperitoneal injection with a potent CYP1A inducer, 3,4,3',4'-tetrachlorobiphenyl (PCB 77). PCB 77 treatment significantly induced hepatic CYP1A activity and protein in yellow bullhead from reference, but not contaminated, sites and had no effect on CYP1A in green sunfish from either site. The low CYP1A expression levels in resident fish with elevated PCB body burdens, together with the failure of PCB injection to induce CYP1A in certain populations, indicate an acclimatory CYP1A response in yellow bullheads and likely an inherently resistant CYP1A in green sunfish. This work demonstrates for the first time acclimation of CYP1A to PCBs in a species within the family Ictaluridae and provides further support for our previous work indicating an apparent inherent lack of CYP1A sensitivity to chlorinated inducers in Centrarchids. These traits may explain, at least in part, the common association of these families with degraded habitats and indicate Lepomis members are likely to be excellent candidates for exploring the mechanistic basis of 'inherent' CYP1A resistance. This study also underlines to the need for thorough characterization of the CYP1A responsivity of a population and/or species prior to using CYP1A as a reliable biomonitoring tool.