Merck
  • Taste-guided fractionation and instrumental analysis of hydrophobic compounds in sake.
Taste-guided fractionation and instrumental analysis of hydrophobic compounds in sake.
Bioscience, biotechnology, and biochemistry (2012-07-13)
Katsumi Hashizume, Toshihiko Ito, Misako Shimohashi, Atsufumi Kokita, Tetsuo Tokiwano, Masaki Okuda
ABSTRACT
The taste-active hydrophobic compounds in a charcoal-untreated sake sample were subjected to a taste dilution analysis (TDA). All of the high-TDA factor fractions showed a bitter or astringent taste in common, but their taste characters were different. The taste-active compounds of the high-TDA factor fractions were purified by taste-guided fractionation, using RP-HPLC and an instrumental analysis. From each of the seven fractions, ferulic acid, ethyl ferulate, tryptophol, three previously reported bitter-tasting peptides, and two novel ethyl esters of the peptides of 10 amino acid residues were identified. All the identified compounds had a similar taste character to that of the TDA fractions analyzed. Ethyl ferulate and the ethyl ester of the peptides showed a moderately bitter taste. The concentration of the identified compounds in seven jyunmai-type sake samples was determined. This concentration was decreased dose dependently by a charcoal treatment which is commonly applied in the final step of sake manufacture, notably with the compounds of high hydrophobicity.
Sigma-Aldrich
3-(2-Hydroxyethyl)indole, 97%
Sigma-Aldrich
Ethyl 4-hydroxy-3-methoxycinnamate, 98%