Merck
  • Genome sequences of two thermophilic Bacillus licheniformis strains, efficient producers of platform chemical 2,3-butanediol.
Genome sequences of two thermophilic Bacillus licheniformis strains, efficient producers of platform chemical 2,3-butanediol.
Journal of bacteriology (2012-07-21)
Lixiang Li, Fei Su, Yu Wang, Lijie Zhang, Cuicui Liu, Jingwen Li, Cuiqing Ma, Ping Xu
ABSTRACT
Both Bacillus licheniformis strains 10-1-A and 5-2-D are efficient producers of 2,3-butanediol. Here we present 4.3-Mb and 4.2-Mb assemblies of their genomes. The key genes for the regulation and metabolism of 2,3-butanediol production were annotated, which may provide further insights into the molecular mechanism for the production of 2,3-butanediol with high yield and productivity.
Sigma-Aldrich
meso-2,3-Butanediol, 99%
Sigma-Aldrich
(2S,3S)-(+)-2,3-Butanediol, 97%
Sigma-Aldrich
(2R,3R)-(−)-2,3-Butanediol, 97%
Sigma-Aldrich
2,3-Butanediol, 98%