Merck
  • Preparation, structural analysis, and properties of tenoxicam cocrystals.
Preparation, structural analysis, and properties of tenoxicam cocrystals.
International journal of pharmaceutics (2012-07-31)
Jagdishwar R Patel, Robert A Carlton, Thomas E Needham, Clinton O Chichester, Frederick G Vogt
ABSTRACT
Cocrystals of tenoxicam, a non-steroidal anti-inflammatory drug, are screened, prepared, and characterized in this study. Nine tenoxicam cocrystals were identified using solvent-drop grinding (SDG) techniques. Structural characterization was performed using powder X-ray diffraction (PXRD), differential scanning calorimetry, and multinuclear solid-state NMR (SSNMR). Thermal analysis, PXRD, and 1D SSNMR are used to detect solvates and phase mixtures encountered in SDG cocrystal screening. 2D SSNMR methods are then used to confirm cocrystal formation and determine structural aspects for selected cocrystals formed with saccharin, salicylic acid, succinic acid, and glycolic acid in comparison to Forms I and III of tenoxicam. Molecular association is demonstrated using cross-polarization heteronuclear dipolar correlation (CP-HETCOR) methods involving (1)H and (13)C nuclei. Short-range (1)H-(13)C CP-HETCOR and (1)H-(1)H double-quantum interactions between atoms of interest, including those engaged in hydrogen bonding, are used to reveal local aspects of the cocrystal structure. (15)N SSNMR is used to assess ionization state and the potential for zwitterionization in the selected cocrystals. The tenoxicam saccharin cocrystal was found to be similar in structure to a previously-reported cocrystal of piroxicam and saccharin. The four selected cocrystals yielded intrinsic dissolution rates that were similar or reduced relative to tenoxicam Form III.
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
Tenoxicam, European Pharmacopoeia (EP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
Sigma-Aldrich
Tenoxicam, NSAID