Merck
  • Staining of the internal limiting membrane with the use of heavy brilliant blue G.
Staining of the internal limiting membrane with the use of heavy brilliant blue G.
Ophthalmic research (2012-08-29)
David E Pelayes, Ferenc Kuhn, Anibal Martin Folgar, Walter Takahashi, Andres Bastien, P Nascimento Vinicius, Jorge Oscar Zarate
ABSTRACT
Brilliant blue G (BBG) is frequently used in chromovitrectomy to facilitate internal limiting membrane (ILM) peeling. A study was initiated to evaluate if heavy BBG is safe and effective in staining the ILM. We studied 30 eyes, 23 with idiopathic macular holes and 7 of patients with diabetic macular edema. Removal of the ILMs was assisted by heavy BBG staining. In cases with histopathological correlation the ILMs were evaluated with hematoxylin and eosin, Masson's trichrome, periodic acid-Schiff and glial fibrillary acidic protein staining. In addition, immunohistochemistry was also performed using specific antibodies for vimentin, neuron-specific enolase, factor VIII and CD68. Using the Image-Pro Plus software of Media Cybernetics Co. we found an average thickness in ILMs. Of the ILM specimens sent, 19/30 (63.33%) could not be processed properly because of the limited sample material, recognizing only fragments of dispersed fibrillar material. In macular hole ILMs we found an average thickness of 1.3 ± 0.65 µm, and in diabetic macular edema ILMs an average thickness of 6.2 ± 1.4 µm. In heavy BBG-assisted ILM peeling we observed no intraoperative or postoperative complications after a mean follow-up of 12 months. Heavy BBG could be an effective and safe vehicle for staining the ILM.
Sigma-Aldrich
Brilliant Blue G, 250, suitable for microscopy
Sigma-Aldrich
Brilliant Blue G solution, Concentrate
Sigma-Aldrich
Brilliant Blue G, pure