Merck
  • Use of xylitol to enhance the therapeutic efficacy of polymethylmethacrylate-based antibiotic therapy in treatment of chronic osteomyelitis.
Use of xylitol to enhance the therapeutic efficacy of polymethylmethacrylate-based antibiotic therapy in treatment of chronic osteomyelitis.
Antimicrobial agents and chemotherapy (2012-09-06)
Karen E Beenken, Laura Bradney, William Bellamy, Robert A Skinner, Sandra G McLaren, M Johannes Gruenwald, Horace J Spencer, James K Smith, Warren O Haggard, Mark S Smeltzer
ABSTRACT
Using a rabbit model of postsurgical osteomyelitis, we demonstrate that incorporation of xylitol into polymethylmethacrylate (PMMA) bone cement enhances the elution of daptomycin under in vivo conditions. We also demonstrate that this can be correlated with an improved therapeutic outcome in the treatment of a chronic bone infection following surgical debridement.
Product Description
Supelco
Xylitol, analytical standard
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Sigma-Aldrich
Xylitol, ≥99% (GC)