Merck
  • In situ assembly of well-dispersed Au nanoparticles on TiO2/ZnO nanofibers: a three-way synergistic heterostructure with enhanced photocatalytic activity.
In situ assembly of well-dispersed Au nanoparticles on TiO2/ZnO nanofibers: a three-way synergistic heterostructure with enhanced photocatalytic activity.
Journal of hazardous materials (2012-09-15)
Peng Zhang, Changlu Shao, Xinghua Li, Mingyi Zhang, Xin Zhang, Yangyang Sun, Yichun Liu
ABSTRACT
The TiO(2)/ZnO nanofibers embedded by Au nanoparticles (TiO(2)/ZnO/Au NFs) were fabricated by combining the electrospinning technique (for TiO(2)/ZnO nanofibers) and an in situ reduction approach (for Au nanoparticles). X-ray diffraction, scanning electron microscopy, energy-dispersive X-ray spectroscopy, transmission electronmicroscopy, X-ray photoelectron spectroscopy, UV-vis diffuse reflectance spectroscopy and photoluminescence spectroscopy, were used to characterize the as-synthesized nanofibers. The results showed that small Au nanoparticles (Au NPs) were well dispersed on the TiO(2)/ZnO nanofibers (TiO(2)/ZnO NFs). And, the TiO(2)/ZnO/Au nanofibers showed high charge separation efficiency under ultraviolet excitation, as evidenced by photoluminescence spectra. The photocatalytic studies revealed that the TiO(2)/ZnO/Au NFs exhibited enhanced photocatalytic efficiency of photodegradation of Methyl orange (MO) and 4-nitrophenol (4-NP) compared with the pure TiO(2) nanofibers, ZnO nanofibers and TiO(2)/ZnO NFs under ultraviolet excitation, which might be attributed to the high separation efficiency of photogenerated electron-hole pairs based on the photosynergistic effect among the three components of TiO(2), ZnO and Au. And, the TiO(2)/ZnO/Au NFs could be easily separated and recycled due to their one-dimensional nanostructural property.
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Methyl Orange, suitable for microscopy