Merck
  • Unfolding of cytochrome C upon interaction with azobenzene-modified copolymers.
Unfolding of cytochrome C upon interaction with azobenzene-modified copolymers.
Biomacromolecules (2012-09-26)
Jing Sun, Juliette Ruchmann, A Pallier, L Jullien, M Desmadril, Christophe Tribet
ABSTRACT
Hydrophilic or amphiphilic macromolecules are common organic matrices used to encapsulate and protect fragile drugs such as proteins. Polymer cargoes are in addition designed for remote control of protein delivery, upon imparting the macromolecules with stimuli-responsive properties, such as light-triggered polarity switches. The effect of interaction between polymers and proteins on the stability of the proteins is, however, rarely investigated. Here we studied the unfolding/folding equilibrium of cytochrome c (cyt c) under its oxidized or reduced forms, in the presence of various amphiphilic copolymers (by circular dichroism and intrinsic fluorescence measurements). As models of stimuli-responsive amphiphilic chains, we considered poly(acrylic acid) derivatives, modified to contain hydrophobic, light-responsive azobenzene moieties. These copolymers are, thus, capable to develop both ionic (under their sodium forms at pH > 8) and hydrophobic associations with the basic protein cyt c (isoelectric point of 10.0). In aqueous buffer upon increasing urea concentrations, cyt c underwent unfolding, at [urea] of 9-10 M, which was analyzed under the framework of the equilibrium between two states (native-unfolded). In the presence of polymers, the native folding of cyt c was preserved at low concentrations of urea (typically <4M). However, the presence of polymers facilitated unfolding, which occurred at urea concentrations lowered by 2-4 M as compared to unfolding in the absence of polymers (polymer/cyt c ratio of 1:1 g/g). The predominant contribution of coulombic interactions was shown by both the lack of significant impact of the amount of (neutral) azobenzene moieties in the copolymers and the disappearance of destabilization at ionic strength higher than 150 mM. In addition, stability was similar to that of an isolated cyt c, in the presence of a neutral chain bearing acryloyl(oligoethyleneoxide) units instead of the ionized sodium acrylate moieties. DSC measurements showed that in the presence of polymers, cyt c is thermally unfolded in aqueous buffer at temperatures lowered by >20 °C as compared to thermal unfolding in the absence of polymers. Upon exposure to UV light, properties of the polymers chains were perturbed in situ, upon cis/trans isomerization of the azobenzene groups. In polymers displaying a photoresponsive polarity and hydrophobicity switch (conventional azobenzene), the stability of cyt c was not affected by the exposure to light. In contrast, when photoionization occurred (using an hydroxyl-azobenzene whose pK(a) can be photoshifted), unfolding was initiated upon exposure to light. Altogether, these results show that coulombic binding is a predominant driving force that facilitates unfolding in water/urea solutions. In regard to the design of light-responsive systems for protein handling and control of folding, we conclude that remote control of the coulombic interaction upon photoionization of chromophores can be more efficient than the more conventional photomodulation of polarity.
Sigma-Aldrich
Azobenzene, 98%